Assay Details
| Gene Symbol: A4GNT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015132078.2 | XP_014987564.1 | 530 | 108 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

A4GNT| Gene Symbol: A4GNT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015132078.2 | XP_014987564.1 | 530 | 108 | ||
Set Membership: |
|