Assay Details
| Gene Symbol: TENT5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001082391.4 | XP_001082391.1 | 330 | 149 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001082391.4 transcripts.
Set Membership: |
|

TENT5A
Genomic Map