Assay Details
| Gene Symbol: DUSP23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015112822.2 | XP_014968308.1 | 445 | 84 | ||
| GenBank mRNA | JU328191.1 | - | 244 | 84 | ||
| JV717742.1 | - | 224 | 84 | |||
More Information
Set Membership: |
|

DUSP23
Genomic Map