Assay Details
| Gene Symbol: OXSM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001091681.4 | XP_001091681.1 | 1622 | 111 | ||
| XM_015132303.2 | XP_014987789.1 | 1968 | 111 | |||
| XM_028843628.1 | XP_028699461.1 | 1800 | 111 | |||
| XM_028843630.1 | XP_028699463.1 | 1426 | 111 | |||
| GenBank mRNA | JU326874.1 | - | 474 | 111 | ||
More Information
Set Membership: |
|

OXSM
Genomic Map