Assay Details
| Gene Symbol: LOC695959 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015127425.2 | XP_014982911.1 | 903 | 85 | ||
| GenBank mRNA | JU320205.1 | - | 545 | 85 | ||
| JV636541.1 | - | 553 | 85 | |||
More Information
Set Membership: |
|

LOC695959
Genomic Map