Assay Details
| Gene Symbol: YOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001083461.4 | XP_001083461.1 | 505 | 160 | ||
| GenBank mRNA | JU489318.1 | - | 245 | 160 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

YOD1| Gene Symbol: YOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001083461.4 | XP_001083461.1 | 505 | 160 | ||
| GenBank mRNA | JU489318.1 | - | 245 | 160 | ||
Set Membership: |
|