Assay Details
| Gene Symbol: SLCO1B3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032941.1 | NP_001028113.1 | 98 | 79 | ||
| XM_015151097.2 | XP_015006583.1 | 240 | 79 | |||
| XM_028828782.1 | XP_028684615.1 | 212 | 79 | |||
| GenBank mRNA | AY787037.1 | - | 98 | 79 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SLCO1B3
Genomic Map