Assay Details
| Gene Symbol: PRDM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028849361.1 | XP_028705194.1 | 2334 | 120 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRDM9| Gene Symbol: PRDM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028849361.1 | XP_028705194.1 | 2334 | 120 | ||
Set Membership: |
|