Assay Details
Gene Symbol: LRRC4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001114570.3 | XP_001114570.1 | 736 | 138 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001114570.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
