Assay Details
| Gene Symbol: MAGEF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001097423.4 | XP_001097423.2 | 148 | 89 | ||
| GenBank mRNA | JV693049.1 | - | 64 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001097423.4 transcripts.
Set Membership: |
|

MAGEF1
Genomic Map