Assay Details
| Gene Symbol: GPR87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001107153.4 | XP_001107153.1 | 382 | 105 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001107153.4 transcripts.
Set Membership: |
|

GPR87
Genomic Map