Assay Details
| Gene Symbol: DCTPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194491.2 | NP_001181420.1 | 298 | 106 | ||
| GenBank mRNA | JU348374.1 | - | 174 | 106 | ||
| JV043578.1 | - | 253 | 106 | |||
| JV696129.1 | - | 233 | 106 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

DCTPP1
Genomic Map