Assay Details
| Gene Symbol: HOXD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015110416.2 | XP_014965902.1 | 30 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015110416.2 transcripts.
Set Membership: |
|

HOXD1
Genomic Map