Assay Details
| Gene Symbol: ZC2HC1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001098038.3 | XP_001098038.3 | 1558 | 144 | ||
| GenBank mRNA | JV723465.1 | - | 1371 | 144 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZC2HC1C| Gene Symbol: ZC2HC1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001098038.3 | XP_001098038.3 | 1558 | 144 | ||
| GenBank mRNA | JV723465.1 | - | 1371 | 144 | ||
Set Membership: |
|