Assay Details
| Gene Symbol: GPR157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015146983.2 | XP_015002469.2 | 515 | 153 | ||
| XM_015147055.2 | XP_015002541.2 | 471 | 153 | |||
| GenBank mRNA | JV662012.1 | - | 337 | 153 | ||
More Information
Set Membership: |
|

GPR157
Genomic Map