Assay Details
| Gene Symbol: TMEM254 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266368.1 | NP_001253297.1 | 113 | 110 | ||
| XM_028853358.1 | XP_028709191.1 | 139 | 110 | |||
| GenBank mRNA | JU336485.1 | - | 113 | 110 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TMEM254
Genomic Map