Assay Details
| Gene Symbol: TASL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194018.1 | NP_001180947.1 | 398 | 112 | ||
| GenBank mRNA | JU493988.1 | - | 252 | 112 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194018.1 transcripts.
Set Membership: |
|

TASL
Genomic Map