Assay Details
| Gene Symbol: ABHD11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001115218.4 | XP_001115218.2 | 948 | 115 | ||
| XM_028845684.1 | XP_028701517.1 | 770 | 115 | |||
| XM_028845685.1 | XP_028701518.1 | 1071 | 115 | |||
| XM_028845686.1 | XP_028701519.1 | 718 | 115 | |||
| GenBank mRNA | JU349201.1 | - | 860 | 115 | ||
| JV559604.1 | - | 711 | 115 | |||
More Information
Set Membership: |
|

ABHD11
Genomic Map