Assay Details
| Gene Symbol: LOC698874 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028849775.1 | XP_028705608.1 | 1497 | 132 | ||
| XM_028849780.1 | XP_028705613.1 | 3904 | 132 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LOC698874
Genomic Map