Assay Details
| Gene Symbol: GPR61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028830632.1 | XP_028686465.1 | 2324 | 70 | ||
| XM_028830635.1 | XP_028686468.1 | 4392 | 70 | |||
| XM_028830637.1 | XP_028686470.1 | 2217 | 70 | |||
| XM_028830640.1 | XP_028686473.1 | 2217 | 70 | |||
| GenBank mRNA | JV522406.1 | - | 584 | 70 | ||
More Information
Set Membership: |
|

GPR61
Genomic Map