Assay Details
| Gene Symbol: B3GNT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001106314.4 | XP_001106314.1 | 4137 | 117 | ||
| XM_028843885.1 | XP_028699718.1 | 3938 | 117 | |||
| XM_028843887.1 | XP_028699720.1 | 4056 | 117 | |||
| XM_028843888.1 | XP_028699721.1 | 4682 | 117 | |||
| XM_028843889.1 | XP_028699722.1 | 3874 | 117 | |||
More Information
Set Membership: |
|

B3GNT5
Genomic Map