Assay Details
| Gene Symbol: CYSTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015140827.2 | XP_014996313.1 | 204 | 163 | ||
| GenBank mRNA | JU336722.1 | - | 139 | 163 | ||
| JU477503.1 | - | 98 | 163 | |||
| JV698703.1 | - | 166 | 163 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay is in the 5'UTR region of the XM_015140827.2 transcripts.
This assay is in the 5'UTR region of the XM_015140827.2 transcripts.
Set Membership: |
|

CYSTM1
Genomic Map