Assay Details
| Gene Symbol: JAGN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260648.2 | NP_001247577.1 | 228 | 82 | ||
| GenBank mRNA | JU322210.1 | - | 223 | 82 | ||
| JU473571.1 | - | 223 | 82 | |||
| JV044446.1 | - | 230 | 82 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

JAGN1
Genomic Map