Assay Details
| Gene Symbol: CABS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001106452.4 | XP_001106452.2 | 56 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001106452.4 transcripts.
Set Membership: |
|

CABS1
Genomic Map