Assay Details
| Gene Symbol: TP53INP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266580.2 | NP_001253509.1 | 489 | 143 | ||
| XM_015145650.2 | XP_015001136.1 | 499 | 143 | |||
| XM_015145655.2 | XP_015001141.1 | 501 | 143 | |||
| XM_028852170.1 | XP_028708003.1 | 957 | 143 | |||
| XM_028852171.1 | XP_028708004.1 | 1199 | 143 | |||
| XM_028852172.1 | XP_028708005.1 | 1100 | 143 | |||
| GenBank mRNA | JU337174.1 | - | 229 | 143 | ||
| JU477901.1 | - | 509 | 143 | |||
| JV048747.1 | - | 493 | 143 | |||
More Information
Set Membership: |
|

TP53INP1
Genomic Map