Assay Details
| Gene Symbol: TP53INP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266580.2 | NP_001253509.1 | 841 | 95 | ||
| XM_015145650.2 | XP_015001136.1 | 851 | 95 | |||
| XM_028852170.1 | XP_028708003.1 | 1309 | 95 | |||
| XM_028852171.1 | XP_028708004.1 | 1551 | 95 | |||
| GenBank mRNA | JU337174.1 | - | 581 | 95 | ||
| JU477901.1 | - | 861 | 95 | |||
| JV048747.1 | - | 845 | 95 | |||
More Information
Set Membership: |
|

TP53INP1
Genomic Map