Assay Details
| Gene Symbol: ADRA1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114733.1 | NP_001108205.1 | 888 | 146 | ||
| XM_015145001.2 | XP_015000487.1 | 1912 | 146 | |||
| XM_015145002.2 | XP_015000488.1 | 1912 | 146 | |||
| XM_015145004.2 | XP_015000490.1 | 1912 | 146 | |||
| XM_015145005.2 | XP_015000491.1 | 1912 | 146 | |||
| XM_028852260.1 | XP_028708093.1 | 2334 | 146 | |||
| GenBank mRNA | AY513585.1 | - | 888 | 146 | ||
| AY851479.1 | - | 888 | 146 | |||
| AY851481.1 | - | 888 | 146 | |||
| AY851482.1 | - | 888 | 146 | |||
| AY851483.1 | - | 888 | 146 | |||
More Information
Set Membership: |
|

ADRA1A
Genomic Map