Assay Details
| Gene Symbol: GPRIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028849927.1 | XP_028705760.1 | 2948 | 153 | ||
| GenBank mRNA | JU332799.1 | - | 2893 | 153 | ||
| JV532465.1 | - | 1118 | 153 | |||
More Information
Set Membership: |
|

GPRIN1
Genomic Map