Assay Details
| Gene Symbol: FKBP1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266723.1 | NP_001253652.1 | 175 | 96 | ||
| XM_015111806.2 | XP_014967292.1 | 220 | 96 | |||
| GenBank mRNA | JU320307.1 | - | 175 | 96 | ||
| JU321344.1 | - | 175 | 96 | |||
More Information
Set Membership: |
|

FKBP1B
Genomic Map