Assay Details
| Gene Symbol: ACTRT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001112288.3 | XP_001112288.1 | 181 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001112288.3 transcripts.
Set Membership: |
|

ACTRT2
Genomic Map