Assay Details
| Gene Symbol: SLC35A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001086178.4 | XP_001086178.2 | 442 | 121 | ||
| GenBank mRNA | JU332789.1 | - | 120 | 121 | ||
| JU474208.1 | - | 114 | 121 | |||
| JV046107.1 | - | 138 | 121 | |||
| JV641444.1 | - | 120 | 121 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001086178.4 transcripts.
Set Membership: |
|

SLC35A4
Genomic Map