Assay Details
| Gene Symbol: CHST14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001096685.4 | XP_001096685.1 | 213 | 167 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001096685.4 transcripts.
Set Membership: |
|

CHST14
Genomic Map