Assay Details
| Gene Symbol: ZNF483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001105826.4 | XP_001105826.1 | 854 | 138 | ||
| XM_015117106.2 | XP_014972592.2 | 854 | 138 | |||
| XM_015117107.2 | XP_014972593.1 | 854 | 138 | |||
| GenBank mRNA | JU339508.1 | - | 840 | 138 | ||
More Information
Set Membership: |
|

ZNF483
Genomic Map