Assay Details
| Gene Symbol: CABLES1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015121660.2 | XP_014977146.2 | 1131 | 70 | ||
| XM_015121661.2 | XP_014977147.2 | 1131 | 70 | |||
| XM_028837915.1 | XP_028693748.1 | 385 | 70 | |||
| GenBank mRNA | JU326389.1 | - | 344 | 70 | ||
| JV598178.1 | - | 604 | 70 | |||
| JV701071.1 | - | 570 | 70 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028837915.1 transcripts.
Set Membership: |
|

CABLES1
Genomic Map