Assay Details
| Gene Symbol: CDC26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015116992.2 | XP_014972478.1 | 316 | 113 | ||
| XM_015116993.2 | XP_014972479.1 | 314 | 113 | |||
| XM_015116994.2 | XP_014972480.1 | 204 | 113 | |||
| GenBank mRNA | JU321412.1 | - | 311 | 113 | ||
| JU333766.1 | - | 147 | 113 | |||
| JU471009.1 | - | 316 | 113 | |||
| JU471010.1 | - | 312 | 113 | |||
| JV045404.1 | - | 270 | 113 | |||
| JV045405.1 | - | 266 | 113 | |||
| JV669047.1 | - | 322 | 113 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015116992.2, XM_015116993.2, XM_015116994.2 transcripts.
Set Membership: |
|

CDC26
Genomic Map