Assay Details
| Gene Symbol: SOCS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028831618.1 | XP_028687451.1 | 4518 | 175 | ||
| XM_028831619.1 | XP_028687452.1 | 4787 | 175 | |||
| XM_028831620.1 | XP_028687453.1 | 4352 | 175 | |||
| GenBank mRNA | JU337260.1 | - | 4338 | 175 | ||
| JU477967.1 | - | 4350 | 175 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SOCS5
Genomic Map