Assay Details
| Gene Symbol: SDR42E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194527.3 | NP_001181456.1 | 75 | 56 | ||
| GenBank mRNA | JV637606.1 | - | 30 | 56 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194527.3 transcripts.
Set Membership: |
|

SDR42E1
Genomic Map