Assay Details
| Gene Symbol: LIPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001103668.3 | XP_001103668.1 | 1222 | 141 | ||
| XM_028832227.1 | XP_028688060.1 | 1373 | 141 | |||
More Information
Set Membership: |
|

LIPT1
Genomic Map