Assay Details
| Gene Symbol: SPATA2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260649.1 | NP_001247578.1 | 67 | 82 | ||
| GenBank mRNA | JU321195.1 | - | 67 | 82 | ||
| JV567194.1 | - | 63 | 82 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001260649.1 transcripts.
Set Membership: |
|

SPATA2L
Genomic Map