Assay Details
| Gene Symbol: C6H5orf24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194883.2 | NP_001181812.1 | 53 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194883.2 transcripts.
Set Membership: |
|

C6H5orf24
Genomic Map