Assay Details
| Gene Symbol: DCAF4L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198702.1 | NP_001185631.1 | 150 | 139 | ||
| GenBank mRNA | JV635965.1 | - | 99 | 139 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001198702.1 transcripts.
Set Membership: |
|

DCAF4L2
Genomic Map