Assay Details
| Gene Symbol: C1H1orf74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001110409.3 | XP_001110409.1 | 652 | 68 | ||
| XM_015117610.2 | XP_014973096.1 | 652 | 68 | |||
| XM_028852774.1 | XP_028708607.1 | 652 | 68 | |||
| GenBank mRNA | JV046259.1 | - | 122 | 68 | ||
| JV635972.1 | - | 146 | 68 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001110409.3, XM_015117610.2, XM_028852774.1 transcripts.
Set Membership: |
|

C1H1orf74
Genomic Map