Assay Details
| Gene Symbol: BHLHA15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015134350.2 | XP_014989836.1 | 460 | 156 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BHLHA15| Gene Symbol: BHLHA15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015134350.2 | XP_014989836.1 | 460 | 156 | ||
Set Membership: |
|