Assay Details
| Gene Symbol: MRAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001096328.4 | XP_001096328.3 | 346 | 153 | ||
| GenBank mRNA | JV656233.1 | - | 160 | 153 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MRAP| Gene Symbol: MRAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001096328.4 | XP_001096328.3 | 346 | 153 | ||
| GenBank mRNA | JV656233.1 | - | 160 | 153 | ||
Set Membership: |
|