Assay Details
Gene Symbol: LOC704239 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015133040.2 | XP_014988526.1 | 789 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
