Assay Details
| Gene Symbol: PKIG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193428.1 | NP_001180357.1 | 515 | 148 | ||
| XM_015148834.2 | XP_015004320.1 | 587 | 148 | |||
| XM_015148836.2 | XP_015004322.1 | 636 | 148 | |||
| XM_015148839.1 | XP_015004325.1 | 439 | 148 | |||
| XM_015148840.2 | XP_015004326.1 | 370 | 148 | |||
| XM_015148841.2 | XP_015004327.1 | 304 | 148 | |||
| XM_028827344.1 | XP_028683177.1 | 430 | 148 | |||
| GenBank mRNA | EG705143.1 | - | 515 | 148 | ||
| JU321278.1 | - | 350 | 148 | |||
| JU321280.1 | - | 498 | 148 | |||
| JU470850.1 | - | 424 | 148 | |||
| JV045246.1 | - | 424 | 148 | |||
| JV704351.1 | - | 523 | 148 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PKIG
Genomic Map