Assay Details
| Gene Symbol: MAGEB10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193982.1 | NP_001180911.1 | 440 | 119 | ||
| XM_015127119.2 | XP_014982605.1 | 634 | 119 | |||
| GenBank mRNA | JV046233.1 | - | 600 | 119 | ||
| JV642705.1 | - | 620 | 119 | |||
More Information
Set Membership: |
|

MAGEB10
Genomic Map