Assay Details
| Gene Symbol: ACADM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001101274.4 | XP_001101274.1 | 926 | 144 | ||
| XM_028832408.1 | XP_028688241.1 | 926 | 144 | |||
| GenBank mRNA | JU322915.1 | - | 531 | 144 | ||
| JU472359.1 | - | 546 | 144 | |||
| JV045728.1 | - | 577 | 144 | |||
| JV601739.1 | - | 592 | 144 | |||
| JV703890.1 | - | 576 | 144 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ACADM
Genomic Map