Assay Details
| Gene Symbol: AGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199093.1 | NP_001186022.1 | 385 | 132 | ||
| XM_015139521.2 | XP_014995007.2 | 506 | 132 | |||
| XM_015139522.2 | XP_014995008.2 | 506 | 132 | |||
| XM_028848028.1 | XP_028703861.1 | 506 | 132 | |||
| XM_028848029.1 | XP_028703862.1 | 506 | 132 | |||
| GenBank mRNA | CN648070.1 | - | 385 | 132 | ||
| JU323397.1 | - | 421 | 132 | |||
| JU472523.1 | - | 461 | 132 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

AGA
Genomic Map