Assay Details
| Gene Symbol: EMD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001090376.4 | XP_001090376.2 | 490 | 104 | ||
| GenBank mRNA | JU323610.1 | - | 230 | 104 | ||
| JV054190.1 | - | 231 | 104 | |||
| JV459610.1 | - | 139 | 104 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

EMD
Genomic Map